Tisk…

3-Chloro-1H-indazole-5-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR305575-1G OR305575-250MG
APOSOR305575-1GEA 16340 CZK
APOSOR305575-1G APOSOR305575-250MG
3-Chloro-1H-indazole-5-carboxylic acid
3-Chloro-1H-indazole-5-carboxylic acid
Vzorec: C₈H₅ClN₂O₂
MW: 196,591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11499013
CAS číslo: 1031417-73-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR