5-Bromo-6-nitro-1H-indazole 97%
Dodavatel: Apollo Scientific
OR305536-1G
OR305536-5G
APOSOR305536-1GEA
1910
CZK
APOSOR305536-1G
APOSOR305536-5G
5-Bromo-6-nitro-1H-indazole 97%
5-Bromo-6-nitro-1H-indazole
Vzorec:
C₇H₄BrN₃O₂ MW: 242,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09027584 CAS číslo: 71785-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...