Tisk…

6-Bromo-1H-pyrazolo[3,4-b]pyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Bromo-7-azaindazole

OR305400-250MG OR305400-1G
APOSOR305400-250MGEA 2820 CZK
APOSOR305400-250MG APOSOR305400-1G
6-Bromo-1H-pyrazolo[3,4-b]pyridine
6-Bromo-1H-pyrazolo[3,4-b]pyridine
Vzorec: C₆H₄BrN₃
MW: 198,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13178164
CAS číslo: 934560-92-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR