6-Methylnaphthalene-1-carboxylic acid
Dodavatel: Apollo Scientific
OR305313-250MG
OR305313-1G
APOSOR305313-250MGEA
7770
CZK
APOSOR305313-250MG
APOSOR305313-1G
6-Methylnaphthalene-1-carboxylic acid
6-Methylnaphthalene-1-carboxylic acid
Vzorec:
C₁₂H₁₀O₂ MW: 186,207 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00092712 CAS číslo: 6315-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...