Tisk…

2-(2-Bromophenyl)-1H-imidazole

Dodavatel: Apollo Scientific
Varování

OR305175-1G OR305175-250MG OR305175-5G
APOSOR305175-1GEA 857 CZK
APOSOR305175-1G APOSOR305175-250MG APOSOR305175-5G
2-(2-Bromophenyl)-1H-imidazole
2-(2-Bromophenyl)-1H-imidazole
Vzorec: C₉H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD08668745
CAS číslo: 162356-38-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR