Tisk…

1-(Imidazo[1,2-a]pyridin-3-yl)ethanone 95%

Dodavatel: Apollo Scientific
Varování

OR305152-250MG OR305152-1G
APOSOR305152-250MGEA 2150 CZK
APOSOR305152-250MG APOSOR305152-1G
1-(Imidazo[1,2-a]pyridin-3-yl)ethanone 95%
1-(Imidazo[1,2-a]pyridin-3-yl)ethanone
Vzorec: C₉H₈N₂O
MW: 160,173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13177175
CAS číslo: 29096-64-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR