Tisk…

6-Bromo-1-isopropyl-1H-indazole 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Bromo-1-(prop-2-yl)-1H-indazole

OR305136-1G OR305136-250MG
APOSOR305136-1GEA 17360 CZK
APOSOR305136-1G APOSOR305136-250MG
6-Bromo-1-isopropyl-1H-indazole 95%
6-Bromo-1-isopropyl-1H-indazole
Vzorec: C₁₀H₁₁BrN₂
MW: 239,112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25460294
CAS číslo: 1214900-44-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR