Tisk…

5-Chloro-1H-indazole-3-carbaldehyde 95%

Dodavatel: Apollo Scientific
Varování

OR305083-1G OR305083-250MG
APOSOR305083-1GEA 5920 CZK
APOSOR305083-1G APOSOR305083-250MG
5-Chloro-1H-indazole-3-carbaldehyde 95%
5-Chloro-1H-indazole-3-carbaldehyde
Vzorec: C₈H₅ClN₂O
MW: 180,591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781535
CAS číslo: 102735-84-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR