2-Bromo-6-nitro-1H-benzimidazole
Dodavatel: Apollo Scientific
|
Varování
|
OR305027-1G
OR305027-250MG
APOSOR305027-1GEA
30120
CZK
APOSOR305027-1G
APOSOR305027-250MG
2-Bromo-6-nitro-1H-benzimidazole
2-Bromo-6-nitro-1H-benzimidazole
Vzorec:
C₇H₄BrN₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD12911562 CAS číslo: 909776-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...