tert-Butyl 5-bromo-1H-benzimidazole-1-carboxylate 97%
Dodavatel: Apollo Scientific
Synonyms:
tert-Butyl 5-bromo-1H-benzo[d]imidazole-1-carboxylate
OR305020-1G
OR305020-250MG
APOSOR305020-1GEA
6250
CZK
APOSOR305020-1G
APOSOR305020-250MG
tert-Butyl 5-bromo-1H-benzimidazole-1-carboxylate 97%
tert-Butyl 5-bromo-1H-benzimidazole-1-carboxylate
Vzorec:
C₁₂H₁₃BrN₂O₂ MW: 297,148 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13182500 CAS číslo: 942590-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...