3-Methoxy-2-methylbenzeneboronic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-Methoxy-2-methylphenyl)boronic acid, 3-Borono-2-methylanisole, 3-Methoxy-2-methylphenylboronic acid, 2-Borono-6-methoxytoluene
OR305006-1G
APOSOR305006-1GEA
4570
CZK
APOSOR305006-1G
3-Methoxy-2-methylbenzeneboronic acid 95%
3-Methoxy-2-methylbenzeneboronic acid
Vzorec:
C₈H₁₁BO₃ MW: 165,982 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18398348 CAS číslo: 1313617-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...