Tisk…

2-Thia-6-azaspiro[3.3]heptane

Dodavatel: Apollo Scientific

OR304423-1G OR304423-250MG OR304423-500MG
APOSOR304423-1GEA 44310 CZK
APOSOR304423-1G APOSOR304423-250MG APOSOR304423-500MG
2-Thia-6-azaspiro[3.3]heptane
2-Thia-6-azaspiro[3.3]heptane
Vzorec: C₅H₉NS
MW: 115,197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17017542
CAS číslo: 920491-09-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR