(6S)-5-Azaspiro[2.4]heptane-6-carboxylic acid hydrochloride
Dodavatel: Apollo Scientific
OR304412-250MG
OR304412-500MG
APOSOR304412-250MGEA
22490
CZK
APOSOR304412-250MG
APOSOR304412-500MG
(6S)-5-Azaspiro[2.4]heptane-6-carboxylic acid hydrochloride
(6S)-5-Azaspiro[2.4]heptane-6-carboxylic acid hydrochloride
Vzorec:
C₇H₁₁NO₂·ClH MW: 177,629 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29918531 CAS číslo: 2200278-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...