1-Methyl-3-oxocyclobutane-1-carbonitrile
Dodavatel: Apollo Scientific
Synonyms:
1-Methyl-3-oxocyclobutanecarbonitrile
OR304361-100MG
OR304361-1G
APOSOR304361-100MGEA
3970
CZK
APOSOR304361-100MG
APOSOR304361-1G
1-Methyl-3-oxocyclobutane-1-carbonitrile
1-Methyl-3-oxocyclobutane-1-carbonitrile
Vzorec:
C₆H₇NO MW: 109,126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22628145 CAS číslo: 32082-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...