cis-3-(Methoxymethyl)cyclobutan-1-amine hydrochloride
Dodavatel: Apollo Scientific
OR304352-1G
OR304352-5G
APOSOR304352-1GEA
16770
CZK
APOSOR304352-1G
APOSOR304352-5G
cis-3-(Methoxymethyl)cyclobutan-1-amine hydrochloride
cis-3-(Methoxymethyl)cyclobutan-1-amine hydrochloride
Vzorec:
C₆H₁₃NO·ClH MW: 151.634 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29918566 CAS číslo: 1068160-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...