Methyl trans-3-bromocyclobutane-1-carboxylate
Dodavatel: Apollo Scientific
Synonyms:
trans-Methyl 3-bromocyclobutanecarboxylate
OR304341-1G
OR304341-250MG
APOSOR304341-1GEA
4630
CZK
APOSOR304341-1G
APOSOR304341-250MG
Methyl trans-3-bromocyclobutane-1-carboxylate
Methyl trans-3-bromocyclobutane-1-carboxylate
Vzorec:
C₆H₉BrO₂ MW: 193,038 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27987093 CAS číslo: 1523618-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...