2-Chloro-5-nitroisonicotinic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR303997-1G
OR303997-5G
APOSOR303997-1GEA
622
CZK
APOSOR303997-1G
APOSOR303997-5G
2-Chloro-5-nitroisonicotinic acid 97%
2-Chloro-5-nitroisonicotinic acid
Vzorec:
C₆H₃ClN₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD14584664 CAS číslo: 907545-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...