Tisk…

2-(Benzylamino)-2-methylpropanenitrile

Dodavatel: Apollo Scientific

OR303996-500MG
APOSOR303996-500MGEA 6090 CZK
APOSOR303996-500MG
2-(Benzylamino)-2-methylpropanenitrile
2-(Benzylamino)-2-methylpropanenitrile
Vzorec: C₁₁H₁₄N₂
MW: 174,242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10014472
CAS číslo: 99840-51-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR