Tisk…

Imidazo[1,2-a]pyridine-7-carbonitrile 98%

Dodavatel: Apollo Scientific

OR303944-1G OR303944-5G
APOSOR303944-1GEA 3110 CZK
APOSOR303944-1G APOSOR303944-5G
Imidazo[1,2-a]pyridine-7-carbonitrile 98%
Imidazo[1,2-a]pyridine-7-carbonitrile
Vzorec: C₈H₅N₃
MW: 143,145 g/mol
Bod tání: 133…134 °C
Storage Temperature: Ambient
MDL Number: MFCD09994633
CAS číslo: 952566-04-0
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR