Tisk…

Diethyl 2-(2-nitrobenzyl)malonate

Dodavatel: Apollo Scientific
Varování

Synonyms: Diethyl 2-(2-nitrobenzyl)propane-1,3-dioate

OR30383-1G OR30383-250MG
APOSOR30383-1GEA 2610 CZK
APOSOR30383-1G APOSOR30383-250MG
Diethyl 2-(2-nitrobenzyl)malonate
Diethyl 2-(2-nitrobenzyl)malonate
Vzorec: C₁₄H₁₇NO₆
Storage Temperature: Ambient
MDL Number: MFCD00110674
CAS číslo: 59803-35-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR