2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Dodavatel: Apollo Scientific
Synonyms:
2,3-Dimethylphenylboronic acid pinacol ester
OR303514-5G
APOSOR303514-5GEA
4900
CZK
APOSOR303514-5G
2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Vzorec:
C₁₄H₂₁BO₂ MW: 232,126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18383605 CAS číslo: 1232132-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...