3-Amino-2,2-dimethyl-3-phenylpropan-1-ol 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR303315-1G
OR303315-500MG
APOSOR303315-1GEA
30290
CZK
APOSOR303315-1G
APOSOR303315-500MG
3-Amino-2,2-dimethyl-3-phenylpropan-1-ol 97%
3-Amino-2,2-dimethyl-3-phenylpropan-1-ol
Vzorec:
C₁₁H₁₇NO MW: 179,259 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18785174 CAS číslo: 156258-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...