Tisk…

3-(3-Chloropropoxy)-4-methoxybenzaldehyde

Dodavatel: Apollo Scientific

OR303312-1G OR303312-500MG
APOSOR303312-1GEA 7670 CZK
APOSOR303312-1G APOSOR303312-500MG
3-(3-Chloropropoxy)-4-methoxybenzaldehyde
3-(3-Chloropropoxy)-4-methoxybenzaldehyde
Vzorec: C₁₁H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD25368746
CAS číslo: 929683-54-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR