Tisk…

3,6-Dibromo-9-butyl-9H-carbazole

Dodavatel: Apollo Scientific
Varování

OR303301-1G OR303301-500MG
APOSOR303301-1GEA 5130 CZK
APOSOR303301-1G APOSOR303301-500MG
3,6-Dibromo-9-butyl-9H-carbazole
3,6-Dibromo-9-butyl-9H-carbazole
Vzorec: C₁₆H₁₅Br₂N
MW: 381.105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22052128
CAS číslo: 121602-03-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR