Methyl 6-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}-2,5-dichloropyrimidine-4-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR303263-500MG
OR303263-1G
APOSOR303263-500MGEA
9560
CZK
APOSOR303263-500MG
APOSOR303263-1G
Methyl 6-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}-2,5-dichloropyrimidine-4-carboxylate
Methyl 6-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}-2,5-dichloropyrimidine-4-carboxylate
Vzorec:
C₁₅H₂₀Cl₂N₄O₄ Bod tání: 123…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD24447578 CAS číslo: 1432896-19-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...