3-(2-Methylpropoxy)-5-(propan-2-yloxy)benzoic acid
Dodavatel: Apollo Scientific
OR303262-1G
OR303262-500MG
OR303262-5G
APOSOR303262-1GEA
9560
CZK
APOSOR303262-1G
APOSOR303262-500MG
APOSOR303262-5G
3-(2-Methylpropoxy)-5-(propan-2-yloxy)benzoic acid
3-(2-Methylpropoxy)-5-(propan-2-yloxy)benzoic acid
Vzorec:
C₁₄H₂₀O₄ Storage Temperature: Ambient |
MDL Number:
MFCD23989321 CAS číslo: 852520-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...