2-Bromo-2-methyl-1-(pyrrolidin-1-yl)propan-1-one
Dodavatel: Apollo Scientific
OR303261-1G
OR303261-500MG
APOSOR303261-1GEA
15310
CZK
APOSOR303261-1G
APOSOR303261-500MG
2-Bromo-2-methyl-1-(pyrrolidin-1-yl)propan-1-one
2-Bromo-2-methyl-1-(pyrrolidin-1-yl)propan-1-one
Vzorec:
C₈H₁₄BrNO Storage Temperature: Ambient |
MDL Number:
MFCD16489085 CAS číslo: 1225773-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...