tert-Butyl N-(2-oxo-2,3-dihydro-1H-indol-6-yl)carbamate
Dodavatel: Apollo Scientific
OR303232-1G
OR303232-500MG
APOSOR303232-1GEA
19110
CZK
APOSOR303232-1G
APOSOR303232-500MG
tert-Butyl N-(2-oxo-2,3-dihydro-1H-indol-6-yl)carbamate
tert-Butyl N-(2-oxo-2,3-dihydro-1H-indol-6-yl)carbamate
Vzorec:
C₁₃H₁₆N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD23697508 CAS číslo: 184021-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...