2-Hydroxy-3-(prop-2-en-1-yl)benzoic acid
Dodavatel: Apollo Scientific
OR303230-1G
OR303230-500MG
APOSOR303230-1GEA
19110
CZK
APOSOR303230-1G
APOSOR303230-500MG
2-Hydroxy-3-(prop-2-en-1-yl)benzoic acid
2-Hydroxy-3-(prop-2-en-1-yl)benzoic acid
Vzorec:
C₁₀H₁₀O₃ MW: 178,185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04110413 CAS číslo: 42729-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...