Tisk…

6-Chloro-1,2,3,4-tetrahydroquinolin-2-one

Dodavatel: Apollo Scientific

Synonyms: 6-Chloro-3,4-dihydroquinolin-2(1H)-one

OR303220-1G OR303220-5G
APOSOR303220-1GEA 1700 CZK
APOSOR303220-1G APOSOR303220-5G
6-Chloro-1,2,3,4-tetrahydroquinolin-2-one
6-Chloro-1,2,3,4-tetrahydroquinolin-2-one
Vzorec: C₉H₈ClNO
MW: 181,619 g/mol
Bod tání: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD09833202
CAS číslo: 19358-40-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR