Dimethyl (morpholin-4-yl)phosphonate
Dodavatel: Apollo Scientific
OR303184-1G
OR303184-500MG
OR303184-5G
APOSOR303184-1GEA
5130
CZK
APOSOR303184-1G
APOSOR303184-500MG
APOSOR303184-5G
Dimethyl (morpholin-4-yl)phosphonate
Dimethyl (morpholin-4-yl)phosphonate
Vzorec:
C₆H₁₄NO₄P MW: 195,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00514647 CAS číslo: 597-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...