Tisk…

3-Chloro-7-methoxyquinoline

Dodavatel: Apollo Scientific

OR303177-1G OR303177-500MG
APOSOR303177-1GEA 15310 CZK
APOSOR303177-1G APOSOR303177-500MG
3-Chloro-7-methoxyquinoline
3-Chloro-7-methoxyquinoline
Vzorec: C₁₀H₈ClNO
MW: 193,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13688991
CAS číslo: 858279-19-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR