Tisk…

Methyl (3E)-4-(3-bromophenyl)-2-oxobut-3-enoate

Dodavatel: Apollo Scientific
Varování

OR303172-1G OR303172-500MG OR303172-5G
APOSOR303172-1GEA 5130 CZK
APOSOR303172-1G APOSOR303172-500MG APOSOR303172-5G
Methyl (3E)-4-(3-bromophenyl)-2-oxobut-3-enoate
Methyl (3E)-4-(3-bromophenyl)-2-oxobut-3-enoate
Vzorec: C₁₁H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD20526360
CAS číslo: 104094-31-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR