Tisk…

2-Bromo-5-(pyrrolidine-1-carbonyl)-1,3-thiazole 97%

Dodavatel: Apollo Scientific

OR303169-1G OR303169-500MG
APOSOR303169-1GEA 13230 CZK
APOSOR303169-1G APOSOR303169-500MG
2-Bromo-5-(pyrrolidine-1-carbonyl)-1,3-thiazole 97%
2-Bromo-5-(pyrrolidine-1-carbonyl)-1,3-thiazole
Vzorec: C₈H₉BrN₂OS
Storage Temperature: Ambient
MDL Number: MFCD20259835
CAS číslo: 877675-06-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR