5-Methoxy-1-benzothiophene-3-carboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR303166-1G
OR303166-500MG
APOSOR303166-1GEA
19110
CZK
APOSOR303166-1G
APOSOR303166-500MG
5-Methoxy-1-benzothiophene-3-carboxylic acid 97%
5-Methoxy-1-benzothiophene-3-carboxylic acid
Vzorec:
C₁₀H₈O₃S Storage Temperature: Ambient |
MDL Number:
MFCD19440790 CAS číslo: 123392-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...