2-(4-Iodopyridin-2-yl)-2-methylpropanenitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR303165-1G
OR303165-500MG
OR303165-5G
APOSOR303165-1GEA
5130
CZK
APOSOR303165-1G
APOSOR303165-500MG
APOSOR303165-5G
2-(4-Iodopyridin-2-yl)-2-methylpropanenitrile
2-(4-Iodopyridin-2-yl)-2-methylpropanenitrile
Vzorec:
C₉H₉IN₂ MW: 272,086 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22575061 CAS číslo: 1217486-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...