Tisk…

4-(Benzyloxy)-3-methylbenzaldehyde

Dodavatel: Apollo Scientific

OR303163-1G OR303163-500MG
APOSOR303163-1GEA 15310 CZK
APOSOR303163-1G APOSOR303163-500MG
4-(Benzyloxy)-3-methylbenzaldehyde
4-(Benzyloxy)-3-methylbenzaldehyde
Vzorec: C₁₅H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD12025073
CAS číslo: 158771-31-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR