Tisk…

Ethyl 6-bromo-1,2-benzothiazole-3-carboxylate

Dodavatel: Apollo Scientific
Varování

OR303160-1G OR303160-250MG OR303160-100MG
APOSOR303160-1GEA 23810 CZK
APOSOR303160-1G APOSOR303160-250MG APOSOR303160-100MG
Ethyl 6-bromo-1,2-benzothiazole-3-carboxylate
Ethyl 6-bromo-1,2-benzothiazole-3-carboxylate
Vzorec: C₁₀H₈BrNO₂S
MW: 286,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11559044
CAS číslo: 858671-74-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR