Methyl (3E)-4-(3-chlorophenyl)-2-oxobut-3-enoate
Dodavatel: Apollo Scientific
OR303158-1G
OR303158-500MG
OR303158-5G
APOSOR303158-1GEA
7670
CZK
APOSOR303158-1G
APOSOR303158-500MG
APOSOR303158-5G
Methyl (3E)-4-(3-chlorophenyl)-2-oxobut-3-enoate
Methyl (3E)-4-(3-chlorophenyl)-2-oxobut-3-enoate
Vzorec:
C₁₁H₉ClO₃ MW: 224,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22689167 CAS číslo: 1112238-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...