Tisk…

1-(Azetidin-3-yl)pyrrolidin-2-one

Dodavatel: Apollo Scientific

OR302762-1G OR302762-250MG
APOSOR302762-1GEA 29560 CZK
APOSOR302762-1G APOSOR302762-250MG
1-(Azetidin-3-yl)pyrrolidin-2-one
1-(Azetidin-3-yl)pyrrolidin-2-one
Vzorec: C₇H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD21643803
CAS číslo: 1257294-05-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR