Piperidine-3-carbonitrile 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR302758-1G
OR302758-5G
APOSOR302758-1GEA
5030
CZK
APOSOR302758-1G
APOSOR302758-5G
Piperidine-3-carbonitrile 95%
Piperidine-3-carbonitrile
Vzorec:
C₆H₁₀N₂ MW: 110,157 g/mol Bod varu: 50 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01321091 CAS číslo: 7492-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...