tert-Butyl 3-aminophenethylcarbamate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl N-[2-(3-aminophenyl)ethyl]carbamate
OR302547-1G
OR302547-250MG
OR302547-5G
APOSOR302547-1GEA
7870
CZK
APOSOR302547-1G
APOSOR302547-250MG
APOSOR302547-5G
tert-Butyl 3-aminophenethylcarbamate
tert-Butyl 3-aminophenethylcarbamate
Vzorec:
C₁₃H₂₀N₂O₂ MW: 236,31 g/mol Bod tání: 103…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD05663960 CAS číslo: 180079-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...