6-Amino-N,N-dimethylpyridine-3-sulphonamide
Dodavatel: Apollo Scientific
OR302536-1G
OR302536-250MG
APOSOR302536-1GEA
22520
CZK
APOSOR302536-1G
APOSOR302536-250MG
6-Amino-N,N-dimethylpyridine-3-sulphonamide
6-Amino-N,N-dimethylpyridine-3-sulphonamide
Vzorec:
C₇H₁₁N₃O₂S MW: 201,246 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06307521 CAS číslo: 627836-23-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...