Tisk…

2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine dihydrochloride

Dodavatel: Apollo Scientific
Varování

OR302510-1G OR302510-250MG
APOSOR302510-1GEA 4770 CZK
APOSOR302510-1G APOSOR302510-250MG
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine dihydrochloride
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine dihydrochloride
Vzorec: C₇H₈N₂·2ClH
Storage Temperature: Ambient
MDL Number: MFCD15144657
CAS číslo: 6000-50-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR