Tisk…

9-Azabicyclo[3.3.1]nonan-3-one

Dodavatel: Apollo Scientific
Nebezpečí

OR302430-1G OR302430-250MG
APOSOR302430-1GEA 10350 CZK
APOSOR302430-1G APOSOR302430-250MG
9-Azabicyclo[3.3.1]nonan-3-one
9-Azabicyclo[3.3.1]nonan-3-one
Vzorec: C₈H₁₃NO
MW: 139,195 g/mol
Bod tání: 123…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02114721
CAS číslo: 4390-39-0
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR