Tisk…

3-(1H-1,2,4-Triazol-1-yl)piperidine 95%

Dodavatel: Apollo Scientific

OR302416-1G OR302416-250MG
APOSOR302416-1GEA 26880 CZK
APOSOR302416-1G APOSOR302416-250MG
3-(1H-1,2,4-Triazol-1-yl)piperidine 95%
3-(1H-1,2,4-Triazol-1-yl)piperidine
Vzorec: C₇H₁₂N₄
MW: 152,197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14632032
CAS číslo: 774511-83-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR