Tisk…

Thiazolo[5,4-c]pyridine-2-thiol 97%

Dodavatel: Apollo Scientific
Varování

OR302398-1G OR302398-100MG OR302398-250MG
APOSOR302398-1GEA 6420 CZK
APOSOR302398-1G APOSOR302398-100MG APOSOR302398-250MG
Thiazolo[5,4-c]pyridine-2-thiol 97%
Thiazolo[5,4-c]pyridine-2-thiol
Vzorec: C₆H₄N₂S₂
MW: 168,239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12755950
CAS číslo: 116990-44-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR