Tisk…

But-3-en-1-amine hydrochloride

Dodavatel: Apollo Scientific
Nebezpečí

OR302284-1G OR302284-5G
APOSOR302284-1GEA 0 CZK
APOSOR302284-1G APOSOR302284-5G
But-3-en-1-amine hydrochloride
But-3-en-1-amine hydrochloride
Vzorec: C₄H₉N·ClH
Bod tání: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD05663670
CAS číslo: 17875-18-2
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR