Tisk…

Ethyl 2-chloroquinazoline-4-carboxylate

Dodavatel: Apollo Scientific
Varování

OR302160-1G OR302160-250MG
APOSOR302160-1GEA 9430 CZK
APOSOR302160-1G APOSOR302160-250MG
Ethyl 2-chloroquinazoline-4-carboxylate
Ethyl 2-chloroquinazoline-4-carboxylate
Vzorec: C₁₁H₉ClN₂O₂
MW: 236,654 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11518957
CAS číslo: 1092352-52-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR