Tisk…

9-Bromo-5,6-dihydro-2-iodoimidazo[1,2-d][1,4]benzoxazepine

Dodavatel: Apollo Scientific

Synonyms: 9-Bromo-2-iodo-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepine

OR302123-1G OR302123-250MG
APOSOR302123-1GEA 12630 CZK
APOSOR302123-1G APOSOR302123-250MG
9-Bromo-5,6-dihydro-2-iodoimidazo[1,2-d][1,4]benzoxazepine
9-Bromo-5,6-dihydro-2-iodoimidazo[1,2-d][1,4]benzoxazepine
Vzorec: C₁₁H₈BrIN₂O
MW: 391,002 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22690733
CAS číslo: 1282516-69-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR